siRNA Details
| Gene Symbol: TMEM220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004313.1 | NP_001004313.1 | 6 | 2748 | ||
| GenBank mRNA | AY550194.1 | - | 6 | 2748 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM220| Gene Symbol: TMEM220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004313.1 | NP_001004313.1 | 6 | 2748 | ||
| GenBank mRNA | AY550194.1 | - | 6 | 2748 | ||