siRNA Details
| Gene Symbol: AGMO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004320.1 | NP_001004320.1 | 1 | 220 | ||
| XM_006715730.1 | XP_006715793.1 | 1 | 220 | |||
| XM_006715731.3 | XP_006715794.1 | 1 | 220 | |||
| XM_011515402.2 | XP_011513704.1 | 1 | 220 | |||
| XM_017012204.1 | XP_016867693.1 | 1 | 220 | |||
| GenBank mRNA | AK131284.1 | - | 1 | 220 | ||
| BC108676.1 | - | 1 | 167 | 6152531 | ||
| KJ534765.1 | - | 1 | 50 | |||

AGMO
Genomic Map