siRNA Details
| Gene Symbol: LOC401261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134654.1 | 1 | 1245 | |||
| GenBank mRNA | AK096036.1 | - | 1 | 1820 | ||
| AK098234.1 | - | 1 | 1245 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC401261| Gene Symbol: LOC401261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134654.1 | 1 | 1245 | |||
| GenBank mRNA | AK096036.1 | - | 1 | 1820 | ||
| AK098234.1 | - | 1 | 1245 | |||