siRNA Details
| Gene Symbol: ZNF705A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004328.2 | NP_001004328.1 | 4 | 367 | ||
| Gene Symbol: ZNF705G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164457.1 | NP_001157929.1 | 4 | 317 | ||
| XM_017012941.1 | XP_016868430.1 | 5 | 450 | |||
| XM_017012942.1 | XP_016868431.1 | 6 | 780 | |||

ZNF705A
Genomic Map