siRNA Details
| Gene Symbol: LINC00238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024338.2 | 4 | 467 | |||
| NR_110307.1 | 3 | 314 | ||||
| NR_110308.1 | 4 | 467 | ||||
| GenBank mRNA | BC033546.1 | - | 4 | 438 | ||
| BC056671.1 | - | 4 | 471 | |||
| BC070365.1 | - | 4 | 447 | |||
| BI829888.1 | - | 3 | 278 | |||

LINC00238
Genomic Map