siRNA Details
| Gene Symbol: BAIAP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026857.1 | 2 | 2720 | |||
| GenBank mRNA | AK027350.1 | - | 1 | 584 | ||
| AK056555.1 | - | 2 | 2702 | |||
| AK075238.1 | - | 1 | 683 | |||
| AK096609.1 | - | 1 | 1628 | |||
| AK124762.1 | - | 2 | 2761 | |||
| BC110822.1 | - | 1 | 729 | |||

BAIAP2-AS1
Genomic Map