siRNA Details
Gene Symbol: STPG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004353.3 | NP_001004353.2 | 6 | 1109 | ||
NM_001256699.1 | NP_001243628.1 | 6 | 1125 | |||
NM_001256700.1 | NP_001243629.1 | 6 | 960 | |||
NM_001256701.1 | NP_001243630.1 | 6 | 944 | |||
NR_046339.1 | 5 | 974 | ||||
NR_046340.1 | 6 | 868 | ||||
XM_006717113.2 | XP_006717176.1 | 7 | 1271 | |||
XM_006717115.2 | XP_006717178.1 | 6 | 951 | |||
GenBank mRNA | AK097565.1 | - | 6 | 1117 | ||
BC033939.1 | - | 6 | 940 | 5272537 | ||
BC050094.1 | - | 5 | 928 | |||
BC132726.1 | - | 6 | 1125 | 40146748 | ||
BC144308.2 | - | 6 | 1109 | 9052828 | ||
BC144311.1 | - | 6 | 944 | 9052831 | ||
BC144314.1 | - | 6 | 1125 | 9052834 |
