siRNA Details
| Gene Symbol: LINGO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004432.3 | NP_001004432.1 | 2 | 2376 | ||
| GenBank mRNA | AY358115.1 | - | 2 | 2343 | ||
| BM716911.1 | - | 1 | 306 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINGO4| Gene Symbol: LINGO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004432.3 | NP_001004432.1 | 2 | 2376 | ||
| GenBank mRNA | AY358115.1 | - | 2 | 2343 | ||
| BM716911.1 | - | 1 | 306 | |||