siRNA Details
| Gene Symbol: OR2D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004684.1 | NP_001004684.1 | 1 | 124 | ||
| GenBank mRNA | BC137208.1 | - | 1 | 152 | 9021205 | |
| BC137209.1 | - | 1 | 152 | 9021206 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2D3| Gene Symbol: OR2D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004684.1 | NP_001004684.1 | 1 | 124 | ||
| GenBank mRNA | BC137208.1 | - | 1 | 152 | 9021205 | |
| BC137209.1 | - | 1 | 152 | 9021206 | ||