siRNA Details
| Gene Symbol: OR2L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004687.1 | NP_001004687.1 | 1 | 51 | ||
| GenBank mRNA | BC137039.1 | - | 1 | 71 | 9021035 | |
| BC137040.1 | - | 1 | 71 | 9021036 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2L3| Gene Symbol: OR2L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004687.1 | NP_001004687.1 | 1 | 51 | ||
| GenBank mRNA | BC137039.1 | - | 1 | 71 | 9021035 | |
| BC137040.1 | - | 1 | 71 | 9021036 | ||