siRNA Details
| Gene Symbol: OR4C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004702.1 | NP_001004702.1 | 1 | 607 | ||
| GenBank mRNA | BC136853.1 | - | 1 | 628 | 9020844 | |
| BC136854.1 | - | 1 | 628 | 9020845 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR4C3| Gene Symbol: OR4C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004702.1 | NP_001004702.1 | 1 | 607 | ||
| GenBank mRNA | BC136853.1 | - | 1 | 628 | 9020844 | |
| BC136854.1 | - | 1 | 628 | 9020845 | ||