siRNA Details
| Gene Symbol: OR5T2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004746.1 | NP_001004746.1 | 1 | 258 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5T2| Gene Symbol: OR5T2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004746.1 | NP_001004746.1 | 1 | 258 | ||
Set Membership: |
|