siRNA Details
| Gene Symbol: KATNAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031303.2 | NP_112593.2 | 4 | 405 | ||
| XM_005258357.4 | XP_005258414.1 | 7 | 950 | |||
| XM_005258358.4 | XP_005258415.1 | 7 | 947 | |||
| XM_005258361.3 | XP_005258418.1 | 3 | 207 | |||
| XM_006722554.3 | XP_006722617.1 | 7 | 1261 | |||
| XM_011526219.2 | XP_011524521.1 | 7 | 950 | |||
| XM_011526220.1 | XP_011524522.1 | 7 | 950 | |||
| XM_011526221.2 | XP_011524523.1 | 7 | 950 | |||
| XM_011526223.2 | XP_011524525.1 | 7 | 950 | |||
| XM_017026028.1 | XP_016881517.1 | 7 | 950 | |||
| XM_017026029.1 | XP_016881518.1 | 7 | 1261 | |||
| XM_017026030.1 | XP_016881519.1 | 6 | 901 | |||
| XM_017026031.1 | XP_016881520.1 | 4 | 386 | |||
| XM_017026032.1 | XP_016881521.1 | 7 | 991 | |||
| GenBank mRNA | BC034999.2 | - | 4 | 405 | 4829800 | |
| CN303333.1 | - | 1 | 108 | |||

KATNAL2
Genomic Map