siRNA Details
| Gene Symbol: FHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033393.2 | NP_203751.2 | 11 | 5380 | ||
| XM_005263319.3 | XP_005263376.1 | 12 | 5521 | |||
| XM_006714393.3 | XP_006714456.1 | 12 | 5530 | |||
| XM_011532388.1 | XP_011530690.1 | 14 | 6012 | |||
| XM_011532389.1 | XP_011530691.1 | 12 | 6126 | |||
| GenBank mRNA | AB051514.1 | - | 11 | 5266 | ||
| AK172734.1 | - | 1 | 834 | |||
| T83061.1 | - | 1 | 223 | |||

FHDC1
Genomic Map