siRNA Details
| Gene Symbol: TP53I13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138349.2 | NP_612358.3 | 6 | 1038 | ||
| XM_017025282.1 | XP_016880771.1 | 6 | 1410 | |||
| GenBank mRNA | AB110209.1 | - | 6 | 1105 | ||
| AK075341.1 | - | 6 | 1069 | |||
| AK093030.1 | - | 1 | 1940 | |||
| AL137404.1 | - | 1 | 308 | |||
| BC001593.2 | - | 6 | 1038 | 2988166 | ||
| BC004507.2 | - | 1 | 410 | |||

TP53I13
Genomic Map