siRNA Details
| Gene Symbol: CARMIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138360.3 | NP_612369.3 | 36 | 3794 | ||
| XM_011537300.2 | XP_011535602.1 | 36 | 3585 | |||
| XM_017021751.1 | XP_016877240.1 | 33 | 3486 | |||
| GenBank mRNA | AK074167.1 | - | 20 | 3517 | ||
| AL834197.1 | - | 7 | 1862 | |||
| BC008134.2 | - | 3 | 241 | |||

CARMIL3
Genomic Map