siRNA Details
| Gene Symbol: H1FNT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181788.1 | NP_861453.1 | 1 | 437 | ||
| GenBank mRNA | AB121028.2 | - | 1 | 77 | ||
| AY302593.1 | - | 1 | 437 | |||
| BC118635.1 | - | 1 | 385 | 40023772 | ||
| BC119787.1 | - | 1 | 387 | 40023773 | ||

H1FNT
Genomic Map