siRNA Details
| Gene Symbol: VMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144939.1 | NP_001138411.1 | 3 | 793 | ||
| NM_001144940.1 | NP_001138412.1 | 3 | 737 | |||
| NM_001144941.1 | NP_001138413.1 | 2 | 641 | |||
| NM_182566.2 | NP_872372.1 | 3 | 757 | |||
| GenBank mRNA | AF521892.1 | - | 3 | 762 | ||
| AY359099.1 | - | 3 | 687 | |||
| BC038467.1 | - | 2 | 640 | |||
| BI836994.1 | - | 2 | 641 | |||

VMO1
Genomic Map