siRNA Details
| Gene Symbol: C16orf74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206967.2 | NP_996850.1 | 4 | 391 | ||
| GenBank mRNA | AB115764.1 | - | 4 | 377 | ||
| AB115765.1 | - | 4 | 389 | |||
| AB115766.1 | - | 2 | 999 | |||
| CD674248.1 | - | 4 | 391 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C16orf74| Gene Symbol: C16orf74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206967.2 | NP_996850.1 | 4 | 391 | ||
| GenBank mRNA | AB115764.1 | - | 4 | 377 | ||
| AB115765.1 | - | 4 | 389 | |||
| AB115766.1 | - | 2 | 999 | |||
| CD674248.1 | - | 4 | 391 | |||