siRNA Details
| Gene Symbol: NAALADL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207015.2 | NP_996898.2 | 10 | 1877 | ||
| XM_006713560.2 | XP_006713623.1 | 12 | 1960 | |||
| XM_011512612.2 | XP_011510914.1 | 10 | 5214 | |||
| XM_011512613.1 | XP_011510915.1 | 10 | 2022 | |||
| XM_011512615.2 | XP_011510917.1 | 9 | 5071 | |||
| XM_011512616.2 | XP_011510918.1 | 9 | 2752 | |||
| XM_011512617.2 | XP_011510919.1 | 10 | 5212 | |||
| XM_017006070.1 | XP_016861559.1 | 14 | 2402 | |||
| XM_017006071.1 | XP_016861560.1 | 15 | 2472 | |||
| XM_017006072.1 | XP_016861561.1 | 16 | 2606 | |||
| XM_017006073.1 | XP_016861562.1 | 14 | 2205 | |||
| XM_017006074.1 | XP_016861563.1 | 16 | 4624 | |||
| XM_017006075.1 | XP_016861564.1 | 13 | 3349 | |||
| XM_017006076.1 | XP_016861565.1 | 13 | 2142 | |||
| XM_017006077.1 | XP_016861566.1 | 15 | 3553 | |||
| XM_017006078.1 | XP_016861567.1 | 13 | 3370 | |||
| XM_017006079.1 | XP_016861568.1 | 11 | 2846 | |||
| XM_017006080.1 | XP_016861569.1 | 11 | 3217 | |||
| XM_017006083.1 | XP_016861572.1 | 10 | 1506 | |||
| GenBank mRNA | AK131044.1 | - | 9 | 1385 | ||
| AL832144.1 | - | 10 | 1877 | |||
| AL832931.1 | - | 7 | 962 | |||

NAALADL2
Genomic Map