siRNA Details
| Gene Symbol: ERICH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207332.2 | NP_997215.1 | 5, 6 | 1346 | ||
| XM_011534731.2 | XP_011533033.1 | 6, 7 | 1518 | |||
| XM_011534739.2 | XP_011533041.1 | 5, 6 | 1325 | |||
| XM_017013123.1 | XP_016868612.1 | 6, 7 | 1533 | |||
| GenBank mRNA | AK290264.1 | - | 5, 6 | 1330 | ||
| BC046243.1 | - | 5, 6 | 1333 | 5207311 | ||
| BX647093.1 | - | 2 | 5675 | |||
| CN353744.1 | - | 1, 2 | 690 | |||

ERICH1
Genomic Map