siRNA Details
| Gene Symbol: PLA2G4F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213600.3 | NP_998765.3 | 20 | 3345 | ||
| NR_033151.1 | 19 | 3272 | ||||
| GenBank mRNA | AK122623.1 | - | 21 | 2956 | ||
| AK172836.1 | - | 14 | 1428 | |||
| AK304368.1 | - | 20 | 3327 | |||
| AL832972.1 | - | 3 | 1319 | |||
| BC035335.1 | - | 19 | 3242 | |||
| BC040920.1 | - | 2 | 1106 | |||
| BC131531.1 | - | 20 | 3257 | 40036909 | ||
| BC142682.1 | - | 20 | 3316 | 40148418 | ||
| BC146648.1 | - | 20 | 3316 | 40147152 | ||
| CR749451.1 | - | 13 | 5418 | |||

PLA2G4F
Genomic Map