siRNA Details
| Gene Symbol: PLA2G4F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213600.3 | NP_998765.3 | 4 | 493 | ||
| NR_033151.1 | 3 | 420 | ||||
| GenBank mRNA | AK122623.1 | - | 4 | 464 | ||
| AK172836.1 | - | 4 | 493 | |||
| AK304368.1 | - | 4 | 475 | |||
| BC035335.1 | - | 3 | 390 | |||
| BC131531.1 | - | 4 | 470 | 40036909 | ||
| BC142682.1 | - | 4 | 464 | 40148418 | ||
| BC146648.1 | - | 4 | 464 | 40147152 | ||

PLA2G4F
Genomic Map