siRNA Details
| Gene Symbol: TMED8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213601.1 | NP_998766.1 | 5 | 637 | ||
| XM_017021223.1 | XP_016876712.1 | 5 | 983 | |||
| XM_017021224.1 | XP_016876713.1 | 4 | 536 | |||
| XM_017021225.1 | XP_016876714.1 | 5 | 3098 | |||
| GenBank mRNA | AK095650.1 | - | 5 | 681 | ||
| AY598329.1 | - | 5 | 637 | |||
| BC101510.1 | - | 5 | 693 | 8069016 | ||
| BX248262.1 | - | 1 | 62 | |||

TMED8
Genomic Map