siRNA Details
| Gene Symbol: OR5M8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005282.1 | NP_001005282.1 | 1 | 126 | ||
| GenBank mRNA | BC136978.1 | - | 1 | 151 | 9020970 | |
| BC136980.1 | - | 1 | 151 | 9020972 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5M8| Gene Symbol: OR5M8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005282.1 | NP_001005282.1 | 1 | 126 | ||
| GenBank mRNA | BC136978.1 | - | 1 | 151 | 9020970 | |
| BC136980.1 | - | 1 | 151 | 9020972 | ||