siRNA Details
| Gene Symbol: OR6K2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005279.1 | NP_001005279.1 | 1 | 582 | ||
| GenBank mRNA | BC137022.1 | - | 1 | 626 | 9021015 | |
| BC137023.1 | - | 1 | 667 | 9021016 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR6K2| Gene Symbol: OR6K2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005279.1 | NP_001005279.1 | 1 | 582 | ||
| GenBank mRNA | BC137022.1 | - | 1 | 626 | 9021015 | |
| BC137023.1 | - | 1 | 667 | 9021016 | ||