siRNA Details
| Gene Symbol: OR8J1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005205.2 | NP_001005205.2 | 1 | 801 | ||
| GenBank mRNA | BC137066.1 | - | 1 | 801 | 9021063 | |
| BC137067.1 | - | 1 | 801 | 9021064 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR8J1| Gene Symbol: OR8J1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005205.2 | NP_001005205.2 | 1 | 801 | ||
| GenBank mRNA | BC137066.1 | - | 1 | 801 | 9021063 | |
| BC137067.1 | - | 1 | 801 | 9021064 | ||