siRNA Details
Gene Symbol: OR52H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005289.1 | NP_001005289.1 | 1 | 318 | ||
GenBank mRNA | BC136998.1 | - | 1 | 344 | 9020990 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR52H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005289.1 | NP_001005289.1 | 1 | 318 | ||
GenBank mRNA | BC136998.1 | - | 1 | 344 | 9020990 |