siRNA Details
| Gene Symbol: OR56A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001917.2 | NP_001001917.2 | 1 | 538 | ||
| GenBank mRNA | BC136904.1 | - | 1 | 538 | 9020895 | |
| BC136911.1 | - | 1 | 538 | 9020902 | ||
More Information
Set Membership: |
|

OR56A1
Genomic Map