siRNA Details
| Gene Symbol: OR8I2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003750.1 | NP_001003750.1 | 1 | 28 | ||
| GenBank mRNA | BC137007.1 | - | 1 | 59 | 9021000 | |
| BC137008.1 | - | 1 | 59 | 9021001 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR8I2| Gene Symbol: OR8I2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003750.1 | NP_001003750.1 | 1 | 28 | ||
| GenBank mRNA | BC137007.1 | - | 1 | 59 | 9021000 | |
| BC137008.1 | - | 1 | 59 | 9021001 | ||