siRNA Details
| Gene Symbol: OTOP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272005.1 | NP_001258934.1 | 2 | 329 | ||
| NM_178233.2 | NP_839947.1 | 2 | 263 | |||
| XM_011524744.2 | XP_011523046.1 | 2 | 259 | |||
| GenBank mRNA | AB593153.1 | - | 2 | 249 | ||
| BC143799.1 | - | 2 | 412 | |||

OTOP3
Genomic Map