siRNA Details
| Gene Symbol: ZNF330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001292002.1 | NP_001278931.1 | 6, 7 | 672 | ||
| NM_014487.5 | NP_055302.1 | 7, 8 | 743 | |||
| XM_017008033.1 | XP_016863522.1 | 7, 8 | 887 | |||
| GenBank mRNA | AJ006591.2 | - | 7, 8 | 688 | ||
| AK002106.1 | - | 7, 8 | 693 | |||
| AK223609.1 | - | 7, 8 | 694 | |||
| AK315463.1 | - | 7, 8 | 729 | |||
| BC004421.1 | - | 7, 8 | 719 | 3352916 | ||

ZNF330
Genomic Map