siRNA Details
| Gene Symbol: EPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB051511.1 | - | 10 | 3851 | ||
| AK023242.1 | - | 1 | 2140 | |||
| AK092710.1 | - | 1 | 2351 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPT1| Gene Symbol: EPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB051511.1 | - | 10 | 3851 | ||
| AK023242.1 | - | 1 | 2140 | |||
| AK092710.1 | - | 1 | 2351 | |||