siRNA Details
| Gene Symbol: ZNF543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213598.3 | NP_998763.2 | 4 | 2772 | ||
| GenBank mRNA | AK131547.1 | - | 4 | 2759 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF543| Gene Symbol: ZNF543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213598.3 | NP_998763.2 | 4 | 2772 | ||
| GenBank mRNA | AK131547.1 | - | 4 | 2759 | ||