siRNA Details
| Gene Symbol: ZSWIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182521.2 | NP_872327.2 | 8 | 1069 | ||
| GenBank mRNA | AK128006.1 | - | 8 | 1069 | ||
| BC031094.1 | - | 8 | 1068 | 5298288 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZSWIM2| Gene Symbol: ZSWIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182521.2 | NP_872327.2 | 8 | 1069 | ||
| GenBank mRNA | AK128006.1 | - | 8 | 1069 | ||
| BC031094.1 | - | 8 | 1068 | 5298288 | ||