siRNA Details
| Gene Symbol: GTPBP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012227.3 | NP_036359.3 | 8 | 1285 | ||
| XM_006724868.3 | XP_006724931.1 | 9 | 1185 | |||
| XM_011545637.2 | XP_011543939.1 | 7 | 890 | |||
| GenBank mRNA | AK024630.1 | - | 8 | 1224 | ||
| AK026462.1 | - | 2 | 306 | |||
| AK074528.1 | - | 2 | 1173 | |||
| AK097872.1 | - | 3 | 1623 | |||
| AK223412.1 | - | 8 | 1230 | |||
| BC014636.1 | - | 8 | 958 | 3528598 | ||
| BT007360.1 | - | 7 | 914 | |||
| Y14391.2 | - | 8 | 1243 | |||

GTPBP6
Genomic Map