siRNA Details
| Gene Symbol: ROM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000327.3 | NP_000318.1 | 3 | 1842 | ||
| GenBank mRNA | BC008100.1 | - | 3 | 1432 | 4177529 | |
| L07894.1 | - | 3 | 1306 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ROM1| Gene Symbol: ROM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000327.3 | NP_000318.1 | 3 | 1842 | ||
| GenBank mRNA | BC008100.1 | - | 3 | 1432 | 4177529 | |
| L07894.1 | - | 3 | 1306 | |||