siRNA Details
| Gene Symbol: CRACR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144958.1 | NP_001138430.1 | 7 | 1185 | ||
| NM_032680.3 | NP_116069.1 | 7 | 1185 | |||
| XM_006719021.3 | XP_006719084.1 | 7 | 1185 | |||
| XM_011521034.2 | XP_011519336.1 | 6 | 932 | |||
| XM_011521036.2 | XP_011519338.1 | 5 | 692 | |||
| GenBank mRNA | AK091124.1 | - | 1 | 21 | ||
| AK304017.1 | - | 7 | 1087 | |||
| BC004524.2 | - | 7 | 1082 | |||
| BC150643.1 | - | 6 | 816 | 9057013 | ||

CRACR2A
Genomic Map