siRNA Details
| Gene Symbol: TMOD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014547.4 | NP_055362.1 | 10 | 1438 | ||
| XM_017022087.1 | XP_016877576.1 | 11 | 1469 | |||
| XM_017022088.1 | XP_016877577.1 | 10 | 1310 | |||
| GenBank mRNA | AF177171.1 | - | 10 | 1400 | ||
| AF237631.1 | - | 9 | 1220 | |||
| AL137543.1 | - | 3 | 367 | |||
| BC020542.2 | - | 10 | 1374 | 4421113 | ||
| BX647277.1 | - | 10 | 1400 | |||

TMOD3
Genomic Map