siRNA Details
| Gene Symbol: PLXNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025179.3 | NP_079455.3 | 2 | 1033 | ||
| XM_005273164.2 | XP_005273221.1 | 2 | 1097 | |||
| XM_005273165.3 | XP_005273222.1 | 2 | 1097 | |||
| GenBank mRNA | AB007932.1 | - | 2 | 484 | ||
| BC132676.1 | - | 2 | 388 | 40146698 | ||
| BC136530.1 | - | 2 | 388 | 9020520 | ||

PLXNA2
Genomic Map