siRNA Details
| Gene Symbol: FAM162A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014367.3 | NP_055182.3 | 4 | 409 | ||
| GenBank mRNA | AF107495.1 | - | 4 | 390 | ||
| AF191020.1 | - | 4 | 372 | |||
| AF201944.1 | - | 4 | 402 | |||
| AF250321.1 | - | 4 | 389 | |||
| AK026030.1 | - | 4 | 403 | |||
| AK125835.1 | - | 1 | 1743 | |||
| AK293994.1 | - | 4 | 387 | |||
| BC010896.1 | - | 4 | 407 | 4283379 | ||
| BC015060.1 | - | 4 | 407 | 4248082 | ||

FAM162A
Genomic Map