siRNA Details
| Gene Symbol: BRWD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007246.2 | NP_001007247.1 | 5 | 557 | ||
| NM_018963.4 | NP_061836.2 | 5 | 557 | |||
| NM_033656.3 | NP_387505.1 | 5 | 557 | |||
| XM_011529611.1 | XP_011527913.1 | 5 | 590 | |||
| XM_017028373.1 | XP_016883862.1 | 6 | 362 | |||
| XM_017028375.1 | XP_016883864.1 | 5 | 590 | |||
| GenBank mRNA | AB080586.1 | - | 5 | 361 | ||
| AB080587.1 | - | 5 | 361 | |||
| AJ238214.2 | - | 5 | 361 | |||
| AJ292465.1 | - | 5 | 341 | |||
| AJ292466.1 | - | 5 | 341 | |||
| BC064602.1 | - | 5 | 393 | |||
| BC142986.1 | - | 5 | 406 | |||

BRWD1
Genomic Map