siRNA Details
| Gene Symbol: SDK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079653.1 | NP_001073121.1 | 16 | 2388 | ||
| NM_152744.3 | NP_689957.3 | 42 | 6243 | |||
| XM_011515188.1 | XP_011513490.1 | 36 | 5084 | |||
| XM_011515189.1 | XP_011513491.1 | 36 | 5081 | |||
| XM_017011837.1 | XP_016867326.1 | 36 | 5084 | |||
| GenBank mRNA | AK055987.1 | - | 3 | 314 | ||
| AK074077.1 | - | 9 | 1183 | |||
| AK074083.1 | - | 27 | 3879 | |||
| AL833354.1 | - | 16 | 2385 | |||
| AY310398.1 | - | 42 | 6243 | |||

SDK1
Genomic Map