siRNA Details
| Gene Symbol: SNX32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152760.2 | NP_689973.2 | 10 | 1016 | ||
| XM_006718488.3 | XP_006718551.1 | 10 | 1319 | |||
| XM_017017470.1 | XP_016872959.1 | 10 | 1294 | |||
| XM_017017471.1 | XP_016872960.1 | 11 | 1478 | |||
| XM_017017472.1 | XP_016872961.1 | 9 | 1680 | |||
| XM_017017473.1 | XP_016872962.1 | 9 | 1680 | |||
| GenBank mRNA | AK055496.1 | - | 8 | 1379 | ||
| AK289587.1 | - | 10 | 982 | |||
| AK299512.1 | - | 8 | 1010 | |||
| AL833261.1 | - | 10 | 1002 | |||
| BC040981.2 | - | 10 | 992 | 4797976 | ||
| BC045563.1 | - | 10 | 965 | 4815203 | ||

SNX32
Genomic Map