siRNA Details
| Gene Symbol: TMEM209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301163.1 | NP_001288092.1 | 5 | 601 | ||
| NM_032842.3 | NP_116231.2 | 5 | 601 | |||
| XM_005250655.1 | XP_005250712.1 | 5 | 891 | |||
| XM_011516646.2 | XP_011514948.1 | 5 | 741 | |||
| GenBank mRNA | AK027678.1 | - | 5 | 506 | ||
| AK027709.1 | - | 5 | 511 | |||
| AK074732.1 | - | 5 | 534 | |||
| BC045618.1 | - | 5 | 509 | 5294911 | ||
| BC057784.1 | - | 5 | 506 | 5313685 | ||
| BU177118.1 | - | 5 | 546 | |||

TMEM209
Genomic Map