siRNA Details
| Gene Symbol: OPRPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021225.4 | NP_067048.4 | 2, 3 | 234 | ||
| GenBank mRNA | CD722615.1 | - | 2, 3 | 234 | ||
| S83198.1 | - | 2, 3 | 140 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OPRPN| Gene Symbol: OPRPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021225.4 | NP_067048.4 | 2, 3 | 234 | ||
| GenBank mRNA | CD722615.1 | - | 2, 3 | 234 | ||
| S83198.1 | - | 2, 3 | 140 | |||