siRNA Details
| Gene Symbol: NKAIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304533.1 | NP_001291462.1 | 3 | 361 | ||
| NR_130764.1 | 3 | 361 | ||||
| XM_011517511.2 | XP_011515813.1 | 3 | 361 | |||
| XM_011517512.2 | XP_011515814.1 | 3 | 361 | |||
| XM_017013359.1 | XP_016868848.1 | 3 | 361 | |||
| XM_017013360.1 | XP_016868849.1 | 3 | 361 | |||
| XM_017013361.1 | XP_016868850.1 | 3 | 1743 | |||
| GenBank mRNA | AK096949.1 | - | 3 | 361 | ||
| AK299689.1 | - | 3 | 634 | |||

NKAIN3
Genomic Map