siRNA Details
| Gene Symbol: RPS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001020.5 | NP_001011.1 | 5 | 574 | ||
| NM_001321111.1 | NP_001308040.1 | 5 | 523 | |||
| XM_005259137.2 | XP_005259194.1 | 4 | 669 | |||
| GenBank mRNA | AK055869.1 | - | 4 | 2347 | ||
| BC004324.2 | - | 5 | 500 | 3628799 | ||
| BC007977.2 | - | 5 | 485 | 4310307 | ||
| BC024970.1 | - | 1 | 470 | |||
| BC071674.1 | - | 4 | 598 | 5394209 | ||
| BC095429.1 | - | 5 | 444 | |||
| BQ421722.1 | - | 5 | 532 | |||
| M60854.1 | - | 5 | 486 | |||

RPS16
Genomic Map