siRNA Details
| Gene Symbol: RPS4Y1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008.3 | NP_000999.1 | 4 | 326 | ||
| XM_017030061.1 | XP_016885550.1 | 4 | 410 | |||
| GenBank mRNA | AK222594.1 | - | 4 | 307 | ||
| AK292819.1 | - | 4 | 311 | |||
| BC010286.1 | - | 3 | 295 | 3048929 | ||
| BC100905.2 | - | 3 | 283 | 40003602 | ||
| BC100906.1 | - | 3 | 281 | |||
| BF242790.1 | - | 4 | 324 | |||
| M58459.1 | - | 3 | 295 | |||

RPS4Y1
Genomic Map