siRNA Details
| Gene Symbol: DCUN1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014283.1 | NP_001014305.1 | 7 | 1297 | ||
| XM_017020643.1 | XP_016876132.1 | 7 | 1508 | |||
| XM_017020644.1 | XP_016876133.1 | 7 | 1438 | |||
| XM_017020645.1 | XP_016876134.1 | 7 | 1479 | |||
| XM_017020646.1 | XP_016876135.1 | 7 | 5220 | |||
| XM_017020647.1 | XP_016876136.1 | 7 | 1758 | |||
| XM_017020648.1 | XP_016876137.1 | 7 | 2521 | |||
| XM_017020649.1 | XP_016876138.1 | 6 | 1335 | |||
| GenBank mRNA | AK000099.1 | - | 5 | 894 | ||
| AK001566.1 | - | 8 | 1432 | |||
| BC056669.1 | - | 7 | 1284 | 6494926 | ||

DCUN1D2
Genomic Map